The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283018.1 | NP_001269947.1 | 3 - 4 | 424 | 72 |
NM_022080.2 | NP_071363.1 | 3 - 4 | 412 | 72 | |
GenBank mRNA | AK124876.1 | - | 3 - 4 | 351 | 72 |
AK294703.1 | - | 2 - 3 | 291 | 72 | |
BC026310.1 | - | 3 - 4 | 359 | 72 | |
BC060840.1 | - | 3 - 4 | 337 | 72 | |
DC335439.1 | - | 3 - 4 | 413 | 72 |