Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPIL4
Entrez Gene ID
85313
Gene Name
peptidylprolyl isomerase like 4
Gene Aliases
HDCME13P
Chromosome Location
Chr.6: 149504495 - 149546102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139126.3 NP_624311.1 2 - 3 231 148
GenBank mRNA AF357880.1 - 2 - 3 164 148
AK092069.1 - 2 - 3 164 148
AK315388.1 - 2 - 3 231 148
BC020986.1 - 2 - 3 167 148
BC064134.1 - 2 - 3 149 148
BC107684.1 - 2 - 3 165 148
BX537536.1 - 2 - 3 165 148

View Full Product Details