The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005603.4 | NP_005594.1 | 15 - 16 | 1752 | 64 |
XM_006722481.3 | XP_006722544.1 | 15 - 16 | 2488 | 64 | |
XM_011526022.2 | XP_011524324.1 | 15 - 16 | 1676 | 64 | |
XM_011526023.2 | XP_011524325.1 | 14 - 15 | 2374 | 64 | |
GenBank mRNA | AF032442.1 | - | 4 - 5 | 471 | 64 |
AF038007.1 | - | 14 - 15 | 1632 | 64 | |
BC003534.1 | - | 4 - 5 | 560 | 64 |