The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005603.4 | NP_005594.1 | 25 - 26 | 3385 | 108 |
XM_006722481.3 | XP_006722544.1 | 25 - 26 | 4121 | 108 | |
XM_011526022.2 | XP_011524324.1 | 25 - 26 | 3309 | 108 | |
XM_011526023.2 | XP_011524325.1 | 24 - 25 | 4007 | 108 | |
GenBank mRNA | AF038007.1 | - | 24 - 25 | 3265 | 108 |
BC003534.1 | - | 14 - 15 | 2193 | 108 |