Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRSF4
Entrez Gene ID
6429
Gene Name
serine and arginine rich splicing factor 4
Gene Aliases
SFRS4, SRP75
Chromosome Location
Chr.1: 29147738 - 29182125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005626.4 NP_005617.2 1 - 2 481 77
XM_011541951.2 XP_011540253.1 1 - 2 281 77
GenBank mRNA AK223444.1 - 1 - 2 258 77
AK291509.1 - 1 - 2 203 77
AK293706.1 - 1 - 2 254 77
BC002781.2 - 1 - 2 205 77
BT007415.1 - 1 - 2 108 77
DC391985.1 - 1 - 2 482 77
L14076.1 - 1 - 2 155 77

View Full Product Details