Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC30A3
Entrez Gene ID
7781
Gene Name
solute carrier family 30 member 3
Gene Aliases
ZNT3
Chromosome Location
Chr.2: 27254572 - 27275863 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318949.1 NP_001305878.1 3 - 4 398 87
NM_001318950.1 NP_001305879.1 3 - 4 543 87
NM_001318951.1 NP_001305880.1 2 - 3 325 87
NM_003459.4 NP_003450.2 2 - 3 461 87
XM_005264547.1 XP_005264604.1 2 - 3 457 87
XM_005264548.3 XP_005264605.1 3 - 4 443 87
XM_006712100.2 XP_006712163.1 2 - 3 1803 87
XM_017004873.1 XP_016860362.1 2 - 3 338 87
XM_017004874.1 XP_016860363.1 3 - 4 586 87
XM_017004875.1 XP_016860364.1 3 - 4 370 87
GenBank mRNA AK302174.1 - 3 - 4 398 87
AK313600.1 - 2 - 3 416 87
AL043136.2 - 2 - 3 461 87
BC028358.2 - 2 - 3 449 87
BM554028.1 - 3 - 4 543 87
DB034101.1 - 2 - 3 325 87
U76010.1 - 2 - 3 358 87

View Full Product Details