The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170795.1 | NP_001164266.1 | 4 - 5 | 386 | 65 |
NM_016085.4 | NP_057169.2 | 4 - 5 | 526 | 65 | |
NM_080592.3 | NP_542159.3 | 4 - 5 | 706 | 65 | |
GenBank mRNA | AB009017.1 | - | 4 - 5 | 399 | 65 |
AF077037.1 | - | 4 - 5 | 388 | 65 | |
AF144055.2 | - | 4 - 5 | 516 | 65 | |
AF275744.1 | - | 5 - 6 | 380 | 65 | |
AK291894.1 | - | 4 - 5 | 516 | 65 | |
AY358968.1 | - | 4 - 5 | 379 | 65 | |
BC002846.1 | - | 4 - 5 | 374 | 65 | |
BC011006.1 | - | 4 - 5 | 380 | 65 | |
BC021237.2 | - | 4 - 5 | 486 | 65 | |
BC035850.1 | - | 4 - 5 | 372 | 65 | |
BQ420456.1 | - | 4 - 5 | 509 | 65 | |
BQ718371.1 | - | 4 - 5 | 421 | 65 | |
CA450289.1 | - | 3 - 4 | 65 | ||
CF126865.1 | - | 4 - 5 | 519 | 65 |