The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003901.3 | NP_003892.2 | 14 - 15 | 1790 | 120 |
XM_005270263.1 | XP_005270320.1 | 14 - 15 | 1803 | 120 | |
XM_006718053.1 | XP_006718116.1 | 15 - 16 | 1905 | 120 | |
XM_011540316.2 | XP_011538618.1 | 14 - 15 | 1849 | 120 | |
XM_011540317.1 | XP_011538619.1 | 14 - 15 | 1699 | 120 | |
XM_011540318.1 | XP_011538620.1 | 13 - 14 | 1720 | 120 | |
GenBank mRNA | AB033078.1 | - | 14 - 15 | 1768 | 120 |
AF144638.1 | - | 13 - 14 | 1568 | 120 | |
AJ011304.2 | - | 14 - 15 | 1745 | 120 | |
AK308610.1 | - | 7 - 8 | 1039 | 120 | |
AK314615.1 | - | 14 - 15 | 1699 | 120 | |
BC052991.1 | - | 14 - 15 | 1778 | 120 |