The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003901.3 | NP_003892.2 | 1 - 2 | 185 | 87 |
XM_006718053.1 | XP_006718116.1 | 1 - 2 | 183 | 87 | |
GenBank mRNA | AB033078.1 | - | 1 - 2 | 163 | 87 |
BC052991.1 | - | 1 - 2 | 173 | 87 | |
BP314104.1 | - | 1 - 2 | 185 | 87 |