Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAPD5
Entrez Gene ID
64282
Gene Name
PAP associated domain containing 5
Gene Aliases
TRF4-2
Chromosome Location
Chr.16: 50152918 - 50235310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040284.2 NP_001035374.2 10 - 11 1599 159
XM_005256092.4 XP_005256149.1 9 - 10 1782 159
XM_005256093.4 XP_005256150.2 10 - 11 1596 159
XM_011523275.2 XP_011521577.1 10 - 11 1595 159
XM_011523278.2 XP_011521580.1 9 - 10 1070 159
XM_017023555.1 XP_016879044.1 10 - 11 1655 159
GenBank mRNA AF089897.1 - 6 - 7 711 159
AK097589.1 - 10 - 11 1427 159
AK223410.1 - 8 - 9 869 159
AK299294.1 - 3 - 4 411 159
AK300918.1 - 10 - 11 1599 159
BC089405.1 - 5 - 6 642 159
BC143706.1 - 5 - 6 616 159
FR872509.1 - 9 - 10 1363 159

View Full Product Details