Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSPAN18
Entrez Gene ID
90139
Gene Name
tetraspanin 18
Gene Aliases
TSPAN
Chromosome Location
Chr.11: 44727148 - 44932427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130783.4 NP_570139.3 6 - 7 671 68
XM_005253217.3 XP_005253274.1 8 - 9 855 68
XM_006718372.3 XP_006718435.1 8 - 9 942 68
XM_006718373.3 XP_006718436.1 7 - 8 928 68
XM_011520459.2 XP_011518761.1 7 - 8 818 68
XM_017018530.1 XP_016874019.1 7 - 8 693 68
XM_017018531.1 XP_016874020.1 6 - 7 615 68
GenBank mRNA AK027715.1 - 5 - 6 480 68
AK075547.1 - 4 - 5 404 68
AY358087.1 - 5 - 6 594 68
BC019342.2 - 4 - 5 443 68

View Full Product Details