Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN18
Entrez Gene ID
90139
Gene Name
tetraspanin 18
Gene Aliases
TSPAN
Chromosome Location
Chr.11: 44727148 - 44932427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130783.4 NP_570139.3 8 - 9 939 100
XM_005253217.3 XP_005253274.1 10 - 11 1123 100
XM_006718372.3 XP_006718435.1 10 - 11 1210 100
XM_006718373.3 XP_006718436.1 9 - 10 1196 100
XM_011520459.2 XP_011518761.1 9 - 10 1086 100
XM_017018530.1 XP_016874019.1 9 - 10 961 100
XM_017018531.1 XP_016874020.1 8 - 9 883 100
GenBank mRNA AK027715.1 - 7 - 8 748 100
AK075547.1 - 6 - 7 672 100
AY358087.1 - 7 - 8 862 100

View Full Product Details