Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AEN
Entrez Gene ID
64782
Gene Name
apoptosis enhancing nuclease
Gene Aliases
ISG20L1, pp12744
Chromosome Location
Chr.15: 88604601 - 88632281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022767.3 NP_073604.3 3 - 4 892 97
XM_005254966.1 XP_005255023.1 3 - 4 1082 97
XM_005254967.1 XP_005255024.1 3 - 4 1222 97
XM_011521905.2 XP_011520207.1 5 - 6 1246 97
XM_017022489.1 XP_016877978.1 3 - 4 1869 97
GenBank mRNA AB209323.1 - 1 - 2 2816 97
AF318330.1 - 3 - 4 822 97
AF327352.1 - 2 - 3 813 97
AK022546.1 - 2 - 3 426 97
BC005164.1 - 2 - 3 808 97
BC014407.1 - 2 - 3 427 97
BC020988.1 - 3 - 4 830 97

View Full Product Details