The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077469.2 | NP_001070937.1 | 1 - 2 | 171 | 94 |
NM_001077471.2 | NP_001070939.1 | 1 - 2 | 171 | 94 | |
NM_001077474.2 | NP_001070942.1 | 1 - 2 | 171 | 94 | |
NM_001077478.2 | NP_001070946.1 | 1 - 2 | 171 | 94 | |
NM_001077480.2 | NP_001070948.1 | 1 - 2 | 171 | 94 | |
NM_001077481.2 | NP_001070949.1 | 1 - 2 | 171 | 94 | |
NM_001077482.2 | NP_001070950.1 | 1 - 2 | 171 | 94 | |
NM_005122.4 | NP_005113.1 | 1 - 2 | 171 | 94 | |
XM_005245697.4 | XP_005245754.1 | 1 - 2 | 54 | 94 | |
GenBank mRNA | AK303796.1 | - | 1 - 2 | 205 | 94 |
AK316208.1 | - | 1 - 2 | 163 | 94 | |
AY572806.1 | - | 1 - 2 | 62 | 94 | |
AY572807.1 | - | 1 - 2 | 62 | 94 | |
AY572810.1 | - | 1 - 2 | 62 | 94 | |
AY572813.1 | - | 1 - 2 | 62 | 94 | |
AY572819.1 | - | 1 - 2 | 62 | 94 | |
AY572822.1 | - | 1 - 2 | 62 | 94 | |
AY572823.1 | - | 1 - 2 | 62 | 94 | |
AY572826.1 | - | 1 - 2 | 62 | 94 | |
BC069626.1 | - | 1 - 2 | 124 | 94 | |
BC121120.1 | - | 1 - 2 | 72 | 94 | |
BC121121.1 | - | 1 - 2 | 72 | 94 | |
CB124867.1 | - | 1 - 2 | 171 | 94 |