Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KATNB1
Entrez Gene ID
10300
Gene Name
katanin regulatory subunit B1
Gene Aliases
KAT, LIS6
Chromosome Location
Chr.16: 57735717 - 57757250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005886.2 NP_005877.2 10 - 11 1231 100
XM_005255772.4 XP_005255829.1 10 - 11 1261 100
XM_006721121.3 XP_006721184.1 10 - 11 1262 100
XM_006721122.2 XP_006721185.1 10 - 11 1386 100
XM_006721123.3 XP_006721186.1 10 - 11 1261 100
XM_011522810.2 XP_011521112.1 10 - 11 1900 100
XM_017022860.1 XP_016878349.1 10 - 11 1203 100
XM_017022861.1 XP_016878350.1 10 - 11 1386 100
XM_017022862.1 XP_016878351.1 10 - 11 1900 100
XM_017022863.1 XP_016878352.1 10 - 11 1400 100
XM_017022864.1 XP_016878353.1 10 - 11 1900 100
GenBank mRNA AF052432.1 - 9 - 10 897 100
BC001353.2 - 9 - 10 880 100
BT007022.1 - 9 - 10 857 100
CR456762.1 - 9 - 10 857 100

View Full Product Details