Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KATNB1
Entrez Gene ID
10300
Gene Name
katanin regulatory subunit B1
Gene Aliases
KAT, LIS6
Chromosome Location
Chr.16: 57735717 - 57757250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006721122.2 XP_006721185.1 1 - 2 260 73
XM_017022861.1 XP_016878350.1 1 - 2 260 73
XM_017022863.1 XP_016878352.1 1 - 2 274 73
GenBank mRNA AB209250.1 - 1 - 2 244 73

View Full Product Details