The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305275.1 | NP_001292204.1 | 18 - 19 | 3299 | 84 |
NM_198576.3 | NP_940978.2 | 18 - 19 | 3299 | 84 | |
XM_005244749.3 | XP_005244806.1 | 18 - 19 | 3320 | 84 | |
XM_011541429.2 | XP_011539731.1 | 18 - 19 | 3320 | 84 | |
GenBank mRNA | AB191264.1 | - | 18 - 19 | 3299 | 84 |
AF016903.1 | - | 18 - 19 | 3192 | 84 | |
BC084578.1 | - | 11 - 12 | 1697 | 84 | |
BF740118.1 | - | 4 - 5 | 84 | ||
CN314678.1 | - | 2 - 3 | 425 | 84 |