The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042454.2 | NP_001035919.1 | 6 - 7 | 558 | 60 |
NM_001164719.1 | NP_001158191.1 | 6 - 7 | 527 | 60 | |
NM_015927.4 | NP_057011.2 | 6 - 7 | 557 | 60 | |
GenBank mRNA | AB007836.1 | - | 5 - 6 | 522 | 60 |
AK122975.1 | - | 4 - 5 | 1881 | 60 | |
AK313327.1 | - | 6 - 7 | 558 | 60 | |
BC001507.3 | - | 6 - 7 | 557 | 60 | |
BC001830.2 | - | 6 - 7 | 557 | 60 | |
BC017288.2 | - | 6 - 7 | 527 | 60 | |
BC032545.1 | - | 5 - 6 | 529 | 60 |