Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KATNB1
Entrez Gene ID
10300
Gene Name
katanin regulatory subunit B1
Gene Aliases
KAT, LIS6
Chromosome Location
Chr.16: 57735717 - 57757250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005886.2 NP_005877.2 1 - 2 111 120
XM_005255772.4 XP_005255829.1 1 - 2 141 120
XM_006721121.3 XP_006721184.1 1 - 2 142 120
XM_006721123.3 XP_006721186.1 1 - 2 141 120
GenBank mRNA BU501165.1 - - 111 120

View Full Product Details