Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT88
Entrez Gene ID
8100
Gene Name
intraflagellar transport 88
Gene Aliases
D13S1056E, DAF19, TG737, TTC10, hTg737
Chromosome Location
Chr.13: 20566446 - 20691437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318491.1 NP_001305420.1 10 - 11 1202 97
NM_001318493.1 NP_001305422.1 12 - 13 1587 97
NM_006531.4 NP_006522.2 11 - 12 1259 97
NM_175605.4 NP_783195.2 13 - 14 1488 97
NR_134653.1 12 - 13 1587 97
XM_005266553.2 XP_005266610.1 13 - 14 1849 97
XM_006719870.3 XP_006719933.1 11 - 12 954 97
XM_011535241.2 XP_011533543.1 12 - 13 1062 97
XM_011535242.1 XP_011533544.1 14 - 15 1989 97
XM_011535243.1 XP_011533545.1 13 - 14 1868 97
XM_017020757.1 XP_016876246.1 12 - 13 1967 97
XM_017020758.1 XP_016876247.1 11 - 12 1463 97
XM_017020759.1 XP_016876248.1 12 - 13 1520 97
XM_017020762.1 XP_016876251.1 12 - 13 1520 97
XM_017020763.1 XP_016876252.1 12 - 13 1520 97
XM_017020765.1 XP_016876254.1 11 - 12 966 97
XM_017020766.1 XP_016876255.1 10 - 11 877 97
XM_017020769.1 XP_016876258.1 10 - 11 889 97
XM_017020774.1 XP_016876263.1 10 - 11 842 97
XM_017020775.1 XP_016876264.1 12 - 13 1520 97
XM_017020776.1 XP_016876265.1 12 - 13 1520 97
GenBank mRNA AK126668.1 - 12 - 13 1958 97
AK300769.1 - 10 - 11 866 97
AK308281.1 - 11 - 12 914 97
AK310223.1 - 13 - 14 1144 97
BC030776.1 - 13 - 14 1169 97
U20362.1 - 11 - 12 1003 97

View Full Product Details