Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT88
Entrez Gene ID
8100
Gene Name
intraflagellar transport 88
Gene Aliases
D13S1056E, DAF19, TG737, TTC10, hTg737
Chromosome Location
Chr.13: 20566446 - 20691437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318491.1 NP_001305420.1 16 - 17 1962 117
NM_001318493.1 NP_001305422.1 18 - 19 2347 117
NM_006531.4 NP_006522.2 17 - 18 2019 117
NM_175605.4 NP_783195.2 19 - 20 2248 117
NR_134653.1 18 - 19 2347 117
XM_005266553.2 XP_005266610.1 19 - 20 2609 117
XM_006719870.3 XP_006719933.1 17 - 18 1714 117
XM_011535241.2 XP_011533543.1 18 - 19 1822 117
XM_011535242.1 XP_011533544.1 20 - 21 2749 117
XM_011535243.1 XP_011533545.1 19 - 20 2628 117
XM_017020757.1 XP_016876246.1 18 - 19 2727 117
XM_017020758.1 XP_016876247.1 17 - 18 2223 117
XM_017020759.1 XP_016876248.1 17 - 18 2193 117
XM_017020760.1 XP_016876249.1 20 - 21 2079 117
XM_017020761.1 XP_016876250.1 19 - 20 1715 117
XM_017020762.1 XP_016876251.1 18 - 19 2280 117
XM_017020763.1 XP_016876252.1 18 - 19 2280 117
XM_017020764.1 XP_016876253.1 18 - 19 1624 117
XM_017020765.1 XP_016876254.1 17 - 18 1726 117
XM_017020766.1 XP_016876255.1 15 - 16 1550 117
XM_017020767.1 XP_016876256.1 17 - 18 1546 117
XM_017020768.1 XP_016876257.1 18 - 19 2091 117
XM_017020769.1 XP_016876258.1 16 - 17 1649 117
XM_017020770.1 XP_016876259.1 20 - 21 2079 117
XM_017020771.1 XP_016876260.1 20 - 21 2507 117
XM_017020772.1 XP_016876261.1 18 - 19 1624 117
XM_017020773.1 XP_016876262.1 19 - 20 2689 117
XM_017020774.1 XP_016876263.1 16 - 17 1602 117
GenBank mRNA AK126668.1 - 18 - 19 2718 117
AK300769.1 - 16 - 17 1626 117
AK310223.1 - 19 - 20 1904 117
BC030776.1 - 19 - 20 1929 117
BX640833.1 - 10 - 11 995 117
U20362.1 - 17 - 18 1763 117

View Full Product Details