Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT88
Entrez Gene ID
8100
Gene Name
intraflagellar transport 88
Gene Aliases
D13S1056E, DAF19, TG737, TTC10, hTg737
Chromosome Location
Chr.13: 20566446 - 20691437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318491.1 NP_001305420.1 18 - 19 2224 143
NM_001318493.1 NP_001305422.1 20 - 21 2609 143
NM_006531.4 NP_006522.2 19 - 20 2281 143
NM_175605.4 NP_783195.2 21 - 22 2510 143
NR_134653.1 20 - 21 2609 143
XM_005266553.2 XP_005266610.1 21 - 22 2871 143
XM_006719870.3 XP_006719933.1 19 - 20 1976 143
XM_011535241.2 XP_011533543.1 20 - 21 2084 143
XM_011535242.1 XP_011533544.1 22 - 23 3011 143
XM_011535243.1 XP_011533545.1 21 - 22 2890 143
XM_017020757.1 XP_016876246.1 20 - 21 2989 143
XM_017020758.1 XP_016876247.1 19 - 20 2485 143
XM_017020759.1 XP_016876248.1 19 - 20 2455 143
XM_017020760.1 XP_016876249.1 22 - 23 2341 143
XM_017020761.1 XP_016876250.1 21 - 22 1977 143
XM_017020762.1 XP_016876251.1 20 - 21 2542 143
XM_017020763.1 XP_016876252.1 20 - 21 2542 143
XM_017020764.1 XP_016876253.1 20 - 21 1886 143
XM_017020765.1 XP_016876254.1 19 - 20 1988 143
XM_017020766.1 XP_016876255.1 17 - 18 1812 143
XM_017020767.1 XP_016876256.1 19 - 20 1808 143
XM_017020768.1 XP_016876257.1 20 - 21 2353 143
XM_017020769.1 XP_016876258.1 18 - 19 1911 143
XM_017020770.1 XP_016876259.1 22 - 23 2341 143
XM_017020771.1 XP_016876260.1 22 - 23 2769 143
XM_017020772.1 XP_016876261.1 20 - 21 1886 143
XM_017020773.1 XP_016876262.1 21 - 22 2951 143
XM_017020774.1 XP_016876263.1 18 - 19 1864 143
GenBank mRNA AK126668.1 - 20 - 21 2980 143
AK300769.1 - 18 - 19 1888 143
AK308281.1 - 19 - 20 1936 143
AK310223.1 - 21 - 22 2166 143
BC030776.1 - 21 - 22 2191 143
U20362.1 - 19 - 20 2025 143

View Full Product Details