Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFAF6
Entrez Gene ID
137682
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 6
Gene Aliases
C8orf38
Chromosome Location
Chr.8: 94895271 - 95118496 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152416.3 NP_689629.2 8 - 9 892 136
XM_005250789.2 XP_005250846.1 8 - 9 903 136
XM_005250790.2 XP_005250847.1 9 - 10 1003 136
XM_005250791.2 XP_005250848.1 9 - 10 969 136
XM_005250792.2 XP_005250849.1 9 - 10 973 136
XM_005250793.2 XP_005250850.1 9 - 10 886 136
XM_011516833.2 XP_011515135.1 13 - 14 2112 136
XM_011516834.2 XP_011515136.1 12 - 13 2073 136
XM_011516835.2 XP_011515137.1 12 - 13 1331 136
XM_011516836.2 XP_011515138.1 12 - 13 2014 136
XM_011516837.2 XP_011515139.1 13 - 14 2072 136
XM_011516838.2 XP_011515140.1 12 - 13 1949 136
XM_011516839.2 XP_011515141.1 11 - 12 1851 136
XM_011516840.2 XP_011515142.1 12 - 13 1945 136
XM_011516841.2 XP_011515143.1 12 - 13 1974 136
XM_011516842.2 XP_011515144.1 11 - 12 1847 136
XM_017013027.1 XP_016868516.1 12 - 13 1323 136
XM_017013028.1 XP_016868517.1 11 - 12 1223 136
XM_017013029.1 XP_016868518.1 10 - 11 1812 136
XM_017013030.1 XP_016868519.1 10 - 11 1557 136
XM_017013031.1 XP_016868520.1 11 - 12 1381 136
XM_017013032.1 XP_016868521.1 9 - 10 1003 136
XM_017013033.1 XP_016868522.1 8 - 9 700 136
XM_017013034.1 XP_016868523.1 7 - 8 673 136
XM_017013035.1 XP_016868524.1 11 - 12 1425 136
XM_017013036.1 XP_016868525.1 11 - 12 1757 136
XM_017013037.1 XP_016868526.1 9 - 10 883 136
XM_017013038.1 XP_016868527.1 10 - 11 952 136
GenBank mRNA AA625387.1 - 4 - 5 356 136
AK298631.1 - 8 - 9 812 136
AY444560.1 - 9 - 10 963 136
BC028166.1 - 10 - 11 1225 136
BF030925.1 - 4 - 5 403 138

View Full Product Details