Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAMKK2
Entrez Gene ID
10645
Gene Name
calcium/calmodulin dependent protein kinase kinase 2
Gene Aliases
CAMKK, CAMKKB
Chromosome Location
Chr.12: 121237692 - 121298308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270485.1 NP_001257414.1 15 - 16 1727 66
NM_001270486.1 NP_001257415.1 14 - 15 1611 66
NM_006549.3 NP_006540.3 15 - 16 2381 66
NM_172214.2 NP_757363.1 15 - 16 2381 66
NM_172215.2 NP_757364.1 14 - 15 2252 66
NM_172216.1 NP_757365.1 14 - 15 2252 66
XM_005253822.3 XP_005253879.1 14 - 15 1554 66
XM_005253823.1 XP_005253880.1 15 - 16 1683 66
XM_011537763.1 XP_011536065.1 15 - 16 1683 66
XM_011537764.2 XP_011536066.1 15 - 16 1729 66
XM_017018694.1 XP_016874183.1 16 - 17 1737 66
XM_017018695.1 XP_016874184.1 16 - 17 1946 66
XM_017018696.1 XP_016874185.1 16 - 17 1788 66
XM_017018697.1 XP_016874186.1 16 - 17 1810 66
XM_017018698.1 XP_016874187.1 15 - 16 1734 66
XM_017018699.1 XP_016874188.1 15 - 16 1608 66
XM_017018701.1 XP_016874190.1 15 - 16 1734 66
XM_017018702.1 XP_016874191.1 15 - 16 1892 66
XM_017018703.1 XP_016874192.1 16 - 17 1737 66
XM_017018704.1 XP_016874193.1 15 - 16 1734 66
XM_017018706.1 XP_016874195.1 15 - 16 1729 66
XM_017018708.1 XP_016874197.1 15 - 16 1729 66
XM_017018709.1 XP_016874198.1 14 - 15 1649 66
GenBank mRNA AB018330.2 - 14 - 15 1557 66
AF091074.1 - 6 - 7 541 66
AF101264.1 - 14 - 15 1204 66
AF140507.1 - 15 - 16 1665 66
AF287630.1 - 15 - 16 2375 66
AF287631.1 - 15 - 16 2375 66
AF321385.1 - 14 - 15 2002 66
AF321386.1 - 14 - 15 2002 66
AL834322.1 - 9 - 10 980 66

View Full Product Details