Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMKK2
Entrez Gene ID
10645
Gene Name
calcium/calmodulin dependent protein kinase kinase 2
Gene Aliases
CAMKK, CAMKKB
Chromosome Location
Chr.12: 121237692 - 121298308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270485.1 NP_001257414.1 6 - 7 934 82
NM_001270486.1 NP_001257415.1 5 - 6 818 82
NM_006549.3 NP_006540.3 6 - 7 1588 82
NM_153499.2 NP_705719.2 6 - 7 1588 82
NM_153500.1 NP_705720.1 6 - 7 1588 82
NM_172214.2 NP_757363.1 6 - 7 1588 82
NM_172215.2 NP_757364.1 6 - 7 1588 82
NM_172216.1 NP_757365.1 6 - 7 1588 82
NM_172226.2 NP_757380.1 6 - 7 934 82
XM_005253822.3 XP_005253879.1 6 - 7 890 82
XM_005253823.1 XP_005253880.1 6 - 7 890 82
XM_005253824.3 XP_005253881.1 6 - 7 890 82
XM_011537763.1 XP_011536065.1 6 - 7 890 82
XM_011537764.2 XP_011536066.1 6 - 7 936 82
XM_017018694.1 XP_016874183.1 7 - 8 944 82
XM_017018695.1 XP_016874184.1 7 - 8 1153 82
XM_017018696.1 XP_016874185.1 7 - 8 995 82
XM_017018697.1 XP_016874186.1 7 - 8 1017 82
XM_017018698.1 XP_016874187.1 6 - 7 941 82
XM_017018699.1 XP_016874188.1 7 - 8 944 82
XM_017018700.1 XP_016874189.1 7 - 8 944 82
XM_017018701.1 XP_016874190.1 6 - 7 941 82
XM_017018702.1 XP_016874191.1 6 - 7 1099 82
XM_017018703.1 XP_016874192.1 7 - 8 944 82
XM_017018704.1 XP_016874193.1 6 - 7 941 82
XM_017018705.1 XP_016874194.1 7 - 8 944 82
XM_017018706.1 XP_016874195.1 6 - 7 936 82
XM_017018707.1 XP_016874196.1 5 - 6 859 82
XM_017018708.1 XP_016874197.1 6 - 7 936 82
GenBank mRNA AB018330.2 - 5 - 6 764 82
AB081336.1 - 5 - 6 791 82
AB081337.1 - 5 - 6 791 82
AF101264.1 - 5 - 6 411 82
AF140507.1 - 6 - 7 872 82
AF287630.1 - 6 - 7 1582 82
AF287631.1 - 6 - 7 1582 82
AF321385.1 - 6 - 7 1338 82
AF321386.1 - 6 - 7 1338 82
AF321387.1 - 6 - 7 1338 82
AF321388.1 - 6 - 7 1338 82
AK024748.1 - 5 - 6 574 82
AK292072.1 - 6 - 7 1082 82
AK300865.1 - 7 - 8 834 82
BC000318.2 - 1 - 2 63 82
BC026060.2 - 6 - 7 867 82

View Full Product Details