Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMKK2
Entrez Gene ID
10645
Gene Name
calcium/calmodulin dependent protein kinase kinase 2
Gene Aliases
CAMKK, CAMKKB
Chromosome Location
Chr.12: 121237692 - 121298308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270485.1 NP_001257414.1 7 - 8 978 59
NM_001270486.1 NP_001257415.1 6 - 7 862 59
NM_006549.3 NP_006540.3 7 - 8 1632 59
NM_153499.2 NP_705719.2 7 - 8 1632 59
NM_153500.1 NP_705720.1 7 - 8 1632 59
NM_172214.2 NP_757363.1 7 - 8 1632 59
NM_172215.2 NP_757364.1 7 - 8 1632 59
NM_172216.1 NP_757365.1 7 - 8 1632 59
NM_172226.2 NP_757380.1 7 - 8 978 59
XM_005253822.3 XP_005253879.1 7 - 8 934 59
XM_005253823.1 XP_005253880.1 7 - 8 934 59
XM_005253824.3 XP_005253881.1 7 - 8 934 59
XM_011537763.1 XP_011536065.1 7 - 8 934 59
XM_011537764.2 XP_011536066.1 7 - 8 980 59
XM_017018694.1 XP_016874183.1 8 - 9 988 59
XM_017018695.1 XP_016874184.1 8 - 9 1197 59
XM_017018696.1 XP_016874185.1 8 - 9 1039 59
XM_017018697.1 XP_016874186.1 8 - 9 1061 59
XM_017018698.1 XP_016874187.1 7 - 8 985 59
XM_017018699.1 XP_016874188.1 8 - 9 988 59
XM_017018700.1 XP_016874189.1 8 - 9 988 59
XM_017018701.1 XP_016874190.1 7 - 8 985 59
XM_017018702.1 XP_016874191.1 7 - 8 1143 59
XM_017018703.1 XP_016874192.1 8 - 9 988 59
XM_017018704.1 XP_016874193.1 7 - 8 985 59
XM_017018705.1 XP_016874194.1 8 - 9 988 59
XM_017018706.1 XP_016874195.1 7 - 8 980 59
XM_017018707.1 XP_016874196.1 6 - 7 903 59
XM_017018708.1 XP_016874197.1 7 - 8 980 59
GenBank mRNA AB018330.2 - 6 - 7 808 59
AB081336.1 - 6 - 7 835 59
AB081337.1 - 6 - 7 835 59
AF101264.1 - 6 - 7 455 59
AF140507.1 - 7 - 8 916 59
AF287630.1 - 7 - 8 1626 59
AF287631.1 - 7 - 8 1626 59
AF321385.1 - 7 - 8 1382 59
AF321386.1 - 7 - 8 1382 59
AF321387.1 - 7 - 8 1382 59
AF321388.1 - 7 - 8 1382 59
AK024748.1 - 6 - 7 618 59
AK300865.1 - 8 - 9 878 59
AL834322.1 - 1 - 2 231 59
BC000318.2 - 2 - 3 107 59
BC026060.2 - 7 - 8 911 59

View Full Product Details