The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123067.3 | NP_001116539.1 | 5 - 6 | 661 | 60 |
NM_001203251.1 | NP_001190180.1 | 5 - 6 | 661 | 60 | |
NM_001203252.1 | NP_001190181.1 | 6 - 7 | 748 | 60 | |
NM_005910.5 | NP_005901.2 | 6 - 7 | 748 | 60 | |
NM_016834.4 | NP_058518.1 | 4 - 5 | 574 | 60 | |
NM_016841.4 | NP_058525.1 | 4 - 5 | 574 | 60 | |
XM_005257367.4 | XP_005257424.1 | 7 - 8 | 1593 | 60 | |
XM_005257368.4 | XP_005257425.1 | 7 - 8 | 1593 | 60 | |
GenBank mRNA | AK095802.1 | - | 4 - 5 | 61 | |
AK226139.1 | - | 2 - 3 | 5711 | 60 | |
AY730549.1 | - | 4 - 5 | 339 | 60 | |
BC000558.2 | - | 4 - 5 | 323 | 60 | |
BC098281.1 | - | 3 - 4 | 272 | 60 | |
BC099721.1 | - | 3 - 4 | 272 | 60 | |
BC101936.1 | - | 3 - 4 | 272 | 60 | |
BC114504.1 | - | 3 - 4 | 272 | 60 | |
BC114948.1 | - | 3 - 4 | 272 | 60 | |
BI552187.1 | - | 5 - 6 | 516 | 60 | |
BT006772.1 | - | 3 - 4 | 252 | 60 | |
J03778.1 | - | 4 - 5 | 289 | 60 | |
X14474.1 | - | 3 - 4 | 282 | 60 |