The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123066.3 | NP_001116538.2 | 8 - 9 | 1699 | 63 |
NM_016835.4 | NP_058519.3 | 8 - 9 | 1699 | 63 | |
XM_005257362.4 | XP_005257419.1 | 8 - 9 | 1791 | 63 | |
XM_005257364.4 | XP_005257421.1 | 7 - 8 | 1704 | 63 | |
XM_005257365.4 | XP_005257422.1 | 8 - 9 | 1791 | 63 | |
XM_005257366.3 | XP_005257423.1 | 6 - 7 | 1659 | 63 | |
XM_005257369.4 | XP_005257426.1 | 7 - 8 | 726 | 63 | |
XM_005257370.4 | XP_005257427.1 | 6 - 7 | 703 | 63 | |
XM_005257371.4 | XP_005257428.1 | 5 - 6 | 552 | 63 | |
GenBank mRNA | AW295014.1 | - | 1 - 2 | 63 |