The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025077.3 | NP_079353.3 | 4 - 5 | 918 | 70 |
XM_005270412.3 | XP_005270469.1 | 4 - 5 | 901 | 70 | |
XM_005270413.4 | XP_005270470.1 | 4 - 5 | 1178 | 70 | |
GenBank mRNA | AK024011.1 | - | 4 - 5 | 439 | 70 |
AK093320.1 | - | 4 - 5 | 605 | 70 | |
AK298162.1 | - | 4 - 5 | 394 | 70 | |
AY169960.1 | - | 4 - 5 | 915 | 70 | |
BC009364.2 | - | 4 - 5 | 352 | 70 | |
CR457320.1 | - | 4 - 5 | 335 | 70 |