The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199415.2 | NP_955447.1 | 3 - 4 | 1448 | 77 |
GenBank mRNA | AK022444.1 | - | 3 - 4 | 1412 | 77 |
BC046122.1 | - | 3 - 4 | 1408 | 77 |