Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CWC25
Entrez Gene ID
54883
Gene Name
CWC25 spliceosome associated protein homolog
Gene Aliases
CCDC49
Chromosome Location
Chr.17: 38800434 - 38825350 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017748.4 NP_060218.1 2 - 3 348 108
GenBank mRNA AK000298.1 - 2 - 3 298 108
AK225373.1 - 2 - 3 319 108
AK302983.1 - 2 - 3 249 108
BC003085.1 - 2 - 3 201 108
BC008833.2 - 2 - 3 334 108
HY021361.1 - 2 - 3 349 108

View Full Product Details