Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AEN
Entrez Gene ID
64782
Gene Name
apoptosis enhancing nuclease
Gene Aliases
ISG20L1, pp12744
Chromosome Location
Chr.15: 88604601 - 88632281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022767.3 NP_073604.3 1 - 2 94 97
XM_006720645.3 XP_006720708.2 1 - 2 53 97
GenBank mRNA AK308119.1 - 1 - 2 55 97

View Full Product Details