Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM2
Entrez Gene ID
2515
Gene Name
ADAM metallopeptidase domain 2
Gene Aliases
CRYN1, CRYN2, CT15, FTNB, PH-30b, PH30, PH30-beta
Chromosome Location
Chr.8: 39743735 - 39838289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278113.1 NP_001265042.1 8 - 9 862 74
NM_001278114.1 NP_001265043.1 9 - 10 919 74
NM_001464.4 NP_001455.3 9 - 10 919 74
XM_005273468.1 XP_005273525.1 9 - 10 857 74
GenBank mRNA AJ133005.1 - 9 - 10 819 74
AK301734.1 - 9 - 10 857 74
BC034957.2 - 8 - 9 782 74
U38805.1 - 9 - 10 883 74
U52370.1 - 9 - 10 822 74
X99374.1 - 9 - 10 815 74

View Full Product Details