The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001232.3 | NP_001223.2 | 1 - 2 | 502 | 110 |
GenBank mRNA | AK129891.1 | - | 1 - 2 | 477 | 110 |
AK225959.1 | - | 1 - 2 | 364 | 110 | |
AK293595.1 | - | 1 - 2 | 511 | 110 | |
AK295502.1 | - | 1 - 2 | 411 | 110 | |
AK313041.1 | - | 1 - 2 | 448 | 110 | |
BC022288.1 | - | 1 - 2 | 411 | 110 | |
D55655.1 | - | 1 - 2 | 347 | 110 |