The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001232.3 | NP_001223.2 | 8 - 9 | 1104 | 71 |
GenBank mRNA | AK129891.1 | - | 8 - 9 | 1079 | 71 |
AK225959.1 | - | 8 - 9 | 966 | 71 | |
AK295502.1 | - | 6 - 7 | 800 | 71 | |
AK313041.1 | - | 8 - 9 | 1050 | 71 | |
BC022288.1 | - | 8 - 9 | 1013 | 71 | |
D55655.1 | - | 8 - 9 | 949 | 71 |