The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136538.1 | NP_001130010.1 | 13 - 14 | 2007 | 67 |
NM_025247.5 | NP_079523.3 | 12 - 13 | 1914 | 67 | |
GenBank mRNA | AK097425.1 | - | 6 - 7 | 864 | 67 |
AL832043.1 | - | 13 - 14 | 1985 | 67 | |
AY323912.1 | - | 12 - 13 | 1870 | 67 | |
BC003698.1 | - | 8 - 9 | 1071 | 67 | |
BC126358.1 | - | 12 - 13 | 1828 | 67 |