The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031215.2 | NP_112492.2 | 1 - 2 | 371 | 81 |
GenBank mRNA | BU166894.1 | - | 1 - 2 | 84 | 81 |
CB960872.1 | - | 1 - 2 | 400 | 82 |