The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163315.2 | NP_001156787.2 | 5 - 6 | 2022 | 82 |
XM_005272048.4 | XP_005272105.1 | 5 - 6 | 2016 | 82 | |
XM_011543574.2 | XP_011541876.1 | 5 - 6 | 2016 | 82 | |
XM_011543575.2 | XP_011541877.1 | 5 - 6 | 2016 | 82 | |
XM_011543576.2 | XP_011541878.1 | 5 - 6 | 2016 | 82 | |
XM_011543577.2 | XP_011541879.1 | 5 - 6 | 2016 | 82 | |
XM_011543578.2 | XP_011541880.1 | 5 - 6 | 2016 | 82 | |
GenBank mRNA | AB533280.1 | - | 6 - 7 | 942 | 82 |
AK126722.1 | - | 6 - 7 | 1210 | 82 | |
AL133602.1 | - | 4 - 5 | 623 | 82 | |
BC126144.1 | - | 5 - 6 | 748 | 82 | |
BC126146.1 | - | 5 - 6 | 748 | 82 |