The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006571.3 | NP_006562.1 | 2 - 3 | 172 | 83 |
GenBank mRNA | AK314128.1 | - | 2 - 3 | 129 | 83 |
BC013175.1 | - | 2 - 3 | 116 | 83 | |
BI562535.1 | - | 2 - 3 | 158 | 83 | |
CR456851.1 | - | 2 - 3 | 85 | 83 | |
D84145.1 | - | 2 - 3 | 172 | 83 |