Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDRG1
Entrez Gene ID
10397
Gene Name
N-myc downstream regulated 1
Gene Aliases
CAP43, CMT4D, DRG-1, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5
Chromosome Location
Chr.8: 133237171 - 133297587 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135242.1 NP_001128714.1 10 - 11 1283 71
NM_001258432.1 NP_001245361.1 8 - 9 769 71
NM_001258433.1 NP_001245362.1 9 - 10 780 71
NM_006096.3 NP_006087.2 10 - 11 886 71
XM_011516791.1 XP_011515093.1 10 - 11 1220 71
XM_011516792.1 XP_011515094.1 3 - 4 1119 71
GenBank mRNA AF004162.1 - 9 - 10 725 71
AF147402.1 - 1 - 2 115 71
AK091147.1 - 9 - 10 729 71
AK095649.1 - 8 - 9 622 71
AK096562.1 - 10 - 11 721 71
AK124709.1 - 2 - 3 1793 71
AK126924.1 - 10 - 11 1565 71
AK223542.1 - 10 - 11 788 71
AK296794.1 - 8 - 9 717 71
AK297342.1 - - 1017 71
AK298236.1 - 8 - 9 687 71
AK307829.1 - 10 - 11 835 71
AK308117.1 - 8 - 9 718 71
AY277597.1 - 9 - 10 714 71
BC003175.1 - 10 - 11 814 71
CR456842.1 - 9 - 10 697 71
D87953.1 - 10 - 11 819 71
X92845.1 - 10 - 11 807 71

View Full Product Details