Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM18
Entrez Gene ID
8749
Gene Name
ADAM metallopeptidase domain 18
Gene Aliases
ADAM27, tMDCIII
Chromosome Location
Chr.8: 39584568 - 39730064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320313.1 NP_001307242.1 11 - 12 1217 112
NM_014237.2 NP_055052.1 12 - 13 1289 112
GenBank mRNA AJ133004.1 - 12 - 13 1234 112
AK313961.1 - 12 - 13 1279 112
AY358321.1 - 11 - 12 1204 112
BC121045.1 - 12 - 13 1234 112

View Full Product Details