Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC1
Entrez Gene ID
58189
Gene Name
WAP four-disulfide core domain 1
Gene Aliases
PS20
Chromosome Location
Chr.16: 84294706 - 84329851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282466.1 NP_001269395.1 1 - 2 406 167
NM_001282467.1 NP_001269396.1 1 - 2 406 167
NM_021197.3 NP_067020.2 1 - 2 406 167
XM_005256079.3 XP_005256136.1 1 - 2 401 167
GenBank mRNA AF169631.1 - 1 - 2 294 167
AF302109.1 - 1 - 2 317 167
BC029159.1 - 1 - 2 212 167
CD519223.1 - 1 - 2 320 167

View Full Product Details