The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017412.3 | NP_059108.1 | 4 - 5 | 915 | 67 |
NM_145866.1 | NP_665873.1 | 3 - 4 | 869 | 67 | |
XM_017013841.1 | XP_016869330.1 | 2 - 3 | 280 | 67 | |
XM_017013842.1 | XP_016869331.1 | 4 - 5 | 905 | 67 | |
XM_017013843.1 | XP_016869332.1 | 4 - 5 | 905 | 67 | |
XM_017013844.1 | XP_016869333.1 | 4 - 5 | 905 | 67 | |
GenBank mRNA | AB039723.1 | - | 4 - 5 | 866 | 67 |
AJ272427.1 | - | 2 - 3 | 722 | 67 | |
AY005130.3 | - | 2 - 3 | 452 | 67 | |
BC034278.1 | - | 2 - 3 | 714 | 67 |