Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
Chromosome Location
Chr.11: 66563463 - 66568576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003793.3 NP_003784.2 11 - 12 1412 66
XM_011545328.2 XP_011543630.1 11 - 12 1162 66
GenBank mRNA AF071748.1 - 9 - 10 976 66
AF071749.1 - 9 - 10 878 66
AF088886.2 - 11 - 12 1412 66
AF136279.1 - 11 - 12 1411 66
AJ007331.1 - 11 - 12 1367 66
AK313657.1 - 11 - 12 1380 66
BC011682.2 - 11 - 12 1406 66
BC036451.1 - 11 - 12 1387 66
CR541680.1 - 10 - 11 884 66

View Full Product Details