Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
Chromosome Location
Chr.11: 66563463 - 66568576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003793.3 NP_003784.2 3 - 4 620 124
XM_011545328.2 XP_011543630.1 3 - 4 370 124
GenBank mRNA AF071748.1 - 1 - 2 184 124
AF071749.1 - 1 - 2 86 124
AF088886.2 - 3 - 4 620 124
AF136279.1 - 3 - 4 619 124
AJ007331.1 - 3 - 4 575 124
AK313657.1 - 3 - 4 588 124
AL137742.1 - 1 - 2 51 124
BC011682.2 - 3 - 4 614 124
BC036451.1 - 3 - 4 595 124
CR541680.1 - 1 - 2 92 124

View Full Product Details