Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
Chromosome Location
Chr.11: 66563463 - 66568576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003793.3 NP_003784.2 8 - 9 1137 62
XM_011545328.2 XP_011543630.1 8 - 9 887 62
GenBank mRNA AF071748.1 - 6 - 7 701 62
AF071749.1 - 6 - 7 603 62
AF088886.2 - 8 - 9 1137 62
AF136279.1 - 8 - 9 1136 62
AJ007331.1 - 8 - 9 1092 62
AK313657.1 - 8 - 9 1105 62
AL137742.1 - 6 - 7 568 62
BC011682.2 - 8 - 9 1131 62
BC036451.1 - 8 - 9 1112 62

View Full Product Details