Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLTSCR2
Entrez Gene ID
29997
Gene Name
glioma tumor suppressor candidate region gene 2
Gene Aliases
PICT-1, PICT1
Chromosome Location
Chr.19: 47745536 - 47757066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015710.4 NP_056525.2 4 - 5 622 122
GenBank mRNA AF182076.1 - 4 - 5 605 122
AK024486.1 - 4 - 5 593 122
AK225915.1 - 4 - 5 651 122
AK297877.1 - 4 - 5 608 122
AK303918.1 - - 609 122
AK309788.1 - 4 - 5 610 122
AL359335.1 - 4 - 5 541 122
AL359336.1 - 4 - 5 565 122
AY535000.1 - 4 - 5 598 122
BC004229.2 - 4 - 5 582 122
BC006311.2 - 4 - 5 604 122
BC007248.1 - 4 - 5 586 122
BC010095.2 - 4 - 5 641 122
BC013203.1 - 4 - 5 610 122
BC013235.1 - 4 - 5 593 122
BC013307.1 - 4 - 5 595 122
BC014009.2 - 4 - 5 592 122
BC039869.1 - 4 - 5 594 122
CX866974.1 - 4 - 5 622 122

View Full Product Details