The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017573.4 | NP_060043.2 | 6 - 7 | 787 | 87 |
XM_011528085.2 | XP_011526387.1 | 7 - 8 | 1012 | 87 | |
XM_011528086.2 | XP_011526388.1 | 7 - 8 | 1012 | 87 | |
XM_011528087.2 | XP_011526389.1 | 7 - 8 | 1012 | 87 | |
XM_011528088.2 | XP_011526390.1 | 7 - 8 | 664 | 87 | |
XM_011528091.2 | XP_011526393.1 | 5 - 6 | 485 | 87 | |
XM_011528092.2 | XP_011526394.1 | 7 - 8 | 1012 | 87 | |
XM_011528093.2 | XP_011526395.1 | 7 - 8 | 1012 | 87 | |
GenBank mRNA | AY358963.1 | - | 6 - 7 | 745 | 87 |