Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCSK4
Entrez Gene ID
54760
Gene Name
proprotein convertase subtilisin/kexin type 4
Gene Aliases
PC4, SPC5
Chromosome Location
Chr.19: 1481428 - 1490874 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017573.4 NP_060043.2 7 - 8 964 88
XM_005259586.1 XP_005259643.1 6 - 7 717 88
XM_011528085.2 XP_011526387.1 8 - 9 1189 88
XM_011528086.2 XP_011526388.1 8 - 9 1189 88
XM_011528087.2 XP_011526389.1 8 - 9 1189 88
XM_011528088.2 XP_011526390.1 8 - 9 841 88
XM_011528091.2 XP_011526393.1 6 - 7 662 88
XM_011528092.2 XP_011526394.1 8 - 9 1189 88
XM_011528093.2 XP_011526395.1 8 - 9 1189 88
XM_011528094.1 XP_011526396.1 5 - 6 796 88
XM_011528095.2 XP_011526397.1 7 - 8 1100 88
XM_017026897.1 XP_016882386.1 4 - 5 477 88
GenBank mRNA AK057235.1 - 5 - 6 1027 88
AY358963.1 - 7 - 8 922 88

View Full Product Details