The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019121.1 | NP_061994.1 | 1 - 2 | 358 | 74 |
XM_006723159.2 | XP_006723222.1 | 1 - 2 | 358 | 74 | |
GenBank mRNA | BC035704.1 | - | 1 - 2 | 580 | 74 |